Assay Details
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007198.3 | NP_009129.1 | 1 - 2 | 179 | 97 | |
XM_017013001.1 | XP_016868490.1 | 1 - 2 | 1009 | 97 | ||
GenBank mRNA | AK291135.1 | - | 1 - 2 | 117 | 97 | |
AL136616.1 | - | 1 - 2 | 159 | 97 | ||
BC012334.1 | - | 1 - 2 | 169 | 3546307 | 97 | |
CR533532.1 | - | 1 - 2 | 102 | 97 | ||
DA086552.1 | - | 1 - 2 | 179 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
