Assay Details
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007198.3 | NP_009129.1 | 5 - 6 | 537 | 112 | |
GenBank mRNA | AK021923.1 | - | 5 - 6 | 405 | 112 | |
AK291135.1 | - | 5 - 6 | 475 | 112 | ||
AL136616.1 | - | 5 - 6 | 517 | 112 | ||
BC012334.1 | - | 5 - 6 | 527 | 3546307 | 112 | |
CR533532.1 | - | 5 - 6 | 460 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
