Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000277.1 | NP_000268.1 | 11 - 12 | 1665 | 101 | |
| GenBank mRNA | AK313383.1 | - | 12 - 13 | 1290 | 101 | |
| BC026251.1 | - | 11 - 12 | 1346 | 4767736 | 101 | |
| K03020.1 | - | 11 - 12 | 1415 | 101 | ||
| L47726.1 | - | 11 - 12 | 1665 | 101 | ||
| U49897.1 | - | 11 - 12 | 1665 | 101 | ||
More Information
Set Membership: |
|

PAH
Genomic Map