Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000277.1 | NP_000268.1 | 1 - 2 | 529 | 72 | |
| XM_017019370.1 | XP_016874859.1 | 1 - 2 | 172 | 72 | ||
| GenBank mRNA | AK298419.1 | - | 1 - 2 | 153 | 72 | |
| AK313383.1 | - | 1 - 2 | 154 | 72 | ||
| BC026251.1 | - | 1 - 2 | 210 | 4767736 | 72 | |
| K03020.1 | - | 1 - 2 | 279 | 72 | ||
| U49897.1 | - | 1 - 2 | 529 | 72 | ||
More Information
Set Membership: |
|

PAH
Genomic Map