Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000277.1 | NP_000268.1 | 3 - 4 | 830 | 74 | |
| XM_017019370.1 | XP_016874859.1 | 3 - 4 | 473 | 74 | ||
| GenBank mRNA | AK298419.1 | - | 3 - 4 | 454 | 74 | |
| AK313383.1 | - | 4 - 5 | 455 | 74 | ||
| BC026251.1 | - | 3 - 4 | 511 | 4767736 | 74 | |
| K03020.1 | - | 3 - 4 | 580 | 74 | ||
| L47726.1 | - | 3 - 4 | 830 | 74 | ||
| U49897.1 | - | 3 - 4 | 830 | 74 | ||
More Information
Set Membership: |
|

PAH
Genomic Map