Assay Details
| Gene Symbol: PTGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000954.5 | NP_000945.3 | 5 - 6 | 627 | 92 | |
| GenBank mRNA | AA621632.1 | - | 2 - 3 | 201 | 92 | |
| AK075333.1 | - | 5 - 6 | 627 | 92 | ||
| BC005939.1 | - | 5 - 6 | 561 | 4294999 | 92 | |
| BC041463.1 | - | 7 - 8 | 1835 | 92 | ||
| M61900.2 | - | 5 - 6 | 617 | 92 | ||
More Information
Set Membership: |
|

PTGDS
Genomic Map