Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177388.1 | NP_001170859.1 | 1 - 2 | 84 | 65 | |
| NM_002306.3 | NP_002297.2 | 1 - 2 | 84 | 65 | ||
| GenBank mRNA | AB006780.1 | - | 1 - 2 | 49 | 65 | |
| AK314929.1 | - | 1 - 2 | 83 | 65 | ||
| BC053667.1 | - | 1 - 2 | 35 | 6148401 | 65 | |
| BC068068.1 | - | 1 - 2 | 30 | 6428428 | 65 | |
| BG332134.1 | - | 1 - 2 | 42 | 65 | ||
| BQ645629.1 | - | 147 | 65 | |||
| CD518618.1 | - | 1 - 2 | 84 | 65 | ||
| M36682.1 | - | 1 - 2 | 29 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB006780.1, AK314929.1, BC053667.1, BC068068.1, M36682.1, NM_001177388.1, NM_002306.3 transcripts.
Set Membership: |
|

LGALS3
Genomic Map