Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177388.1 | NP_001170859.1 | 2 - 3 | 110 | 85 | |
| NM_002306.3 | NP_002297.2 | 2 - 3 | 110 | 85 | ||
| XM_011536759.2 | XP_011535061.2 | 3 - 4 | 100 | 126 | ||
| GenBank mRNA | AB006780.1 | - | 2 - 3 | 75 | 85 | |
| AK314929.1 | - | 2 - 3 | 109 | 85 | ||
| BC001120.2 | - | 2 - 3 | 46 | 3050135 | 85 | |
| BC053667.1 | - | 2 - 3 | 61 | 6148401 | 85 | |
| BC068068.1 | - | 2 - 3 | 56 | 6428428 | 85 | |
| BG332134.1 | - | 2 - 3 | 68 | 85 | ||
| BQ645629.1 | - | 173 | 85 | |||
| CD518618.1 | - | 2 - 3 | 110 | 85 | ||
| M36682.1 | - | 2 - 3 | 55 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB006780.1, AK314929.1, BC001120.2, BC053667.1, BC068068.1, M36682.1, NM_001177388.1, NM_002306.3, XM_011536759.2 transcripts.
Set Membership: |
|

LGALS3
Genomic Map