Assay Details
| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278640.1 | NP_001265569.1 | 1 - 2 | 243 | 104 | |
| NM_002882.3 | NP_002873.1 | 1 - 2 | 243 | 104 | ||
| XM_011530289.2 | XP_011528591.1 | 1 - 2 | 265 | 104 | ||
| XM_011530290.2 | XP_011528592.1 | 1 - 2 | 279 | 104 | ||
| XM_017028890.1 | XP_016884379.1 | 1 - 2 | 265 | 104 | ||
| GenBank mRNA | AK223506.1 | - | 1 - 2 | 162 | 104 | |
| AK293502.1 | - | 1 - 2 | 110 | 104 | ||
| AK300927.1 | - | 1 - 2 | 162 | 104 | ||
| CR456556.1 | - | 1 - 2 | 71 | 104 | ||
| D38076.1 | - | 1 - 2 | 166 | 104 | ||
| DB459798.1 | - | 1 - 2 | 245 | 104 | ||
| X83617.1 | - | 1 - 2 | 119 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293502.1, AK300927.1, CR456556.1, D38076.1, NM_001278640.1, NM_002882.3, X83617.1, XM_011530289.2, XM_011530290.2, XM_017028890.1 transcripts.
Set Membership: |
|

RANBP1
Genomic Map