Assay Details
| Gene Symbol: KLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006563.3 | NP_006554.1 | 1 - 2 | 151 | 116 | |
| GenBank mRNA | BC033580.1 | - | 1 - 2 | 118 | 5201847 | 116 |
| BC040000.1 | - | 1 - 2 | 129 | 116 | ||
| U65404.1 | - | 1 - 2 | 129 | 116 | ||
More Information
Set Membership: |
|

KLF1
Genomic Map