Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006873.3 | NP_006864.2 | 1 - 2 | 53 | 92 | |
| GenBank mRNA | AK314464.1 | - | 1 - 2 | 53 | 92 | |
| DB288942.1 | - | 1 - 2 | 53 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK314464.1, NM_006873.3 transcripts.
Set Membership: |
|

STON1
Genomic Map