Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014079.3 | NP_054798.1 | 1 - 2 | 155 | 139 | |
| GenBank mRNA | BC036733.1 | - | 1 - 2 | 167 | 5526657 | 139 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC036733.1, NM_014079.3 transcripts.
Set Membership: |
|

KLF15
Genomic Map