Assay Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040410.2 | NP_001035500.1 | 1 - 2 | 171 | 80 | |
| GenBank mRNA | DB091135.1 | - | 1 - 2 | 166 | 80 | |
More Information
Set Membership: |
|

C9orf131
Genomic Map