Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080441.1 | NP_001073910.1 | 1 - 2 | 115 | 67 | |
| GenBank mRNA | BC146916.1 | - | 1 - 2 | 138 | 9056844 | 67 |
| BC146921.1 | - | 1 - 2 | 138 | 9056849 | 67 | |
| EU489483.1 | - | 1 - 2 | 177 | 67 | ||
More Information
Set Membership: |
|

TTC36
Genomic Map