Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016093.3 | NP_057177.1 | 1 - 2 | 44 | 75 | |
| XM_005265921.3 | XP_005265978.1 | 1 - 2 | 95 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_016093.3 transcripts.
Set Membership: |
|

RPL26L1
Genomic Map