Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018357.3 | NP_060827.2 | 1 - 2 | 375 | 62 | |
| GenBank mRNA | AF443828.1 | - | 1 - 2 | 268 | 62 | |
| AF443829.1 | - | 1 - 2 | 268 | 62 | ||
| AL833876.1 | - | 1 - 2 | 229 | 62 | ||
| BC006082.1 | - | 1 - 2 | 222 | 2961587 | 62 | |
| BC009446.1 | - | 1 - 2 | 222 | 3529516 | 62 | |
| BC014018.2 | - | 1 - 2 | 226 | 4127153 | 62 | |
| BI913258.1 | - | 1 - 2 | 283 | 62 | ||
| DN992782.1 | - | 1 - 2 | 380 | 62 | ||
More Information
Set Membership: |
|

LARP6
Genomic Map