Assay Details
| Gene Symbol: MAML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018717.4 | NP_061187.2 | 4 - 5 | 3149 | 73 | |
| GenBank mRNA | AK123604.1 | - | 4 - 5 | 2467 | 73 | |
| AK296891.1 | - | 3 - 4 | 441 | 73 | ||
| BC137130.1 | - | 4 - 5 | 2462 | 9021127 | 73 | |
| BC137131.1 | - | 4 - 5 | 2462 | 9021128 | 73 | |
More Information
Set Membership: |
|

MAML3
Genomic Map