Assay Details
| Gene Symbol: NMUR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020167.4 | NP_064552.3 | 3 - 4 | 1102 | 100 | |
| GenBank mRNA | AB041228.1 | - | 3 - 4 | 936 | 100 | |
| AF242874.1 | - | 3 - 4 | 927 | 100 | ||
| AF272363.1 | - | 3 - 4 | 962 | 100 | ||
| AF292402.1 | - | 3 - 4 | 936 | 100 | ||
| BC016938.2 | - | 3 - 4 | 1085 | 3852151 | 100 | |
| BC067776.1 | - | 3 - 4 | 1102 | 30336834 | 100 | |
More Information
Set Membership: |
|

NMUR2
Genomic Map