Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020345.3 | NP_065078.1 | 2 - 3 | 379 | 85 | |
| XM_005265076.3 | XP_005265133.1 | 2 - 3 | 266 | 85 | ||
| XM_005265078.3 | XP_005265135.1 | 2 - 3 | 218 | 85 | ||
| GenBank mRNA | AK027749.1 | - | 2 - 3 | 356 | 85 | |
| BC012145.1 | - | 2 - 3 | 262 | 4549097 | 85 | |
| BG713995.1 | - | 2 - 3 | 384 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK027749.1, BC012145.1, NM_020345.3, XM_005265076.3, XM_005265078.3 transcripts.
Set Membership: |
|

NKIRAS1
Genomic Map