Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020345.3 | NP_065078.1 | 4 - 5 | 740 | 99 | |
| XM_005265076.3 | XP_005265133.1 | 4 - 5 | 627 | 99 | ||
| XM_005265077.4 | XP_005265134.1 | 3 - 4 | 595 | 99 | ||
| XM_005265078.3 | XP_005265135.1 | 4 - 5 | 579 | 99 | ||
| GenBank mRNA | AF229839.1 | - | 2 - 3 | 338 | 99 | |
| BC012145.1 | - | 4 - 5 | 623 | 4549097 | 99 | |
| BC066940.1 | - | 4 - 5 | 631 | 5273965 | 99 | |
More Information
Set Membership: |
|

NKIRAS1
Genomic Map