Assay Details
| Gene Symbol: CNTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024877.3 | NP_079153.2 | 2 - 3 | 407 | 77 | |
| XM_006723395.3 | XP_006723458.2 | 2 - 3 | 828 | 77 | ||
| GenBank mRNA | AK301833.1 | - | 2 - 3 | 407 | 77 | |
More Information
Set Membership: |
|

CNTD2
Genomic Map