Assay Details
| Gene Symbol: CCDC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030771.1 | NP_110398.1 | 5 - 6 | 962 | 94 | |
| GenBank mRNA | AF301222.1 | - | 2 - 3 | 205 | 94 | |
| AF382034.1 | - | 5 - 6 | 849 | 94 | ||
| BC029421.1 | - | 5 - 6 | 635 | 94 | ||
More Information
Set Membership: |
|

CCDC34
Genomic Map