Assay Details
Gene Symbol: OR2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030908.2 | NP_112170.1 | 1 | 1715 | 105 | |
GenBank mRNA | BF848491.1 | - | 1 | 257 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
