Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031923.3 | NP_114129.1 | 4 - 5 | 2524 | 91 | |
| XM_011519741.1 | XP_011518043.1 | 4 - 5 | 2548 | 91 | ||
More Information
Set Membership: |
|

TAF3
Genomic Map