Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031923.3 | NP_114129.1 | 6 - 7 | 2880 | 59 | |
| XM_011519741.1 | XP_011518043.1 | 6 - 7 | 2904 | 59 | ||
More Information
Set Membership: |
|

TAF3
Genomic Map