Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032377.3 | NP_115753.1 | 1 - 2 | 48 | 64 | |
| GenBank mRNA | AK097850.1 | - | 1 - 2 | 42 | 64 | |
| BG718042.1 | - | 1 - 2 | 152 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_032377.3 transcripts.
Set Membership: |
|

ELOF1
Genomic Map