Assay Details
| Gene Symbol: USP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267169.3 | XP_005267226.1 | 1 - 2 | 129 | 129 | |
| XM_005267172.3 | XP_005267229.1 | 1 - 2 | 98 | 129 | ||
| XM_005267175.3 | XP_005267232.1 | 1 - 2 | 97 | 129 | ||
| XM_017011383.1 | XP_016866872.1 | 1 - 2 | 49 | 129 | ||
| XM_017011385.1 | XP_016866874.1 | 1 - 2 | 97 | 129 | ||
| XM_017011386.1 | XP_016866875.1 | 1 - 2 | 97 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267169.3, XM_005267172.3, XM_005267175.3, XM_017011383.1, XM_017011385.1, XM_017011386.1 transcripts.
Set Membership: |
|

USP45
Genomic Map