Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080626.5 | NP_542193.3 | 1 - 2 | 357 | 72 | |
| XM_011537940.2 | XP_011536242.1 | 1 - 2 | 345 | 72 | ||
| GenBank mRNA | AF283671.2 | - | 1 - 2 | 96 | 72 | |
| AF284094.1 | - | 1 - 2 | 301 | 72 | ||
| BC034525.1 | - | 1 - 2 | 334 | 4778946 | 72 | |
| CA488612.1 | - | 368 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF283671.2 transcripts.
Set Membership: |
|

BRI3BP
Genomic Map