Assay Details
| Gene Symbol: CST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130794.1 | NP_570612.1 | 1 - 2 | 260 | 86 | |
| GenBank mRNA | AF335480.1 | - | 1 - 2 | 260 | 86 | |
| BC121080.2 | - | 1 - 2 | 257 | 40119722 | 86 | |
More Information
Set Membership: |
|

CST11
Genomic Map