Assay Details
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138378.2 | NP_612387.1 | 2 - 3 | 656 | 65 | |
GenBank mRNA | BC006136.1 | - | 2 - 3 | 335 | 65 | |
BC127680.1 | - | 2 - 3 | 379 | 40127680 | 65 | |
CN260853.1 | - | 2 - 3 | 363 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
