Assay Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138395.3 | NP_612404.1 | 1 | 1618 | 60 | |
| GenBank mRNA | AB107013.1 | - | 1 | 1561 | 60 | |
| AK098121.1 | - | 1 | 1553 | 60 | ||
| AK301252.1 | - | 2 | 1333 | 60 | ||
| BC009115.1 | - | 1 | 1521 | 60 | ||
| BC040934.2 | - | 1 | 596 | 60 | ||
| BC126294.1 | - | 1 | 1556 | 8992010 | 60 | |
More Information
Set Membership: |
|

MARS2
Genomic Map