Assay Details
| Gene Symbol: C6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145029.3 | NP_659466.2 | 1 - 2 | 263 | 55 | |
| GenBank mRNA | BM695485.1 | - | 1 - 2 | 138 | 55 | |
More Information
Set Membership: |
|

C6orf136
Genomic Map