Assay Details
| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000304.3 | NP_000295.1 | 2 - 3 | 318 | 73 | |
| NM_001281455.1 | NP_001268384.1 | 2 - 3 | 279 | 73 | ||
| NM_001281456.1 | NP_001268385.1 | 2 - 3 | 314 | 73 | ||
| NM_153321.2 | NP_696996.1 | 2 - 3 | 275 | 73 | ||
| NM_153322.2 | NP_696997.1 | 1 - 2 | 197 | 73 | ||
| XM_017024775.1 | XP_016880264.1 | 1 - 2 | 114 | 73 | ||
| XM_017024776.1 | XP_016880265.1 | 2 - 3 | 287 | 73 | ||
| GenBank mRNA | AK290640.1 | - | 2 - 3 | 228 | 73 | |
| AK300690.1 | - | 2 - 3 | 170 | 73 | ||
| BC019040.2 | - | 2 - 3 | 240 | 4588473 | 73 | |
| BC091499.1 | - | 2 - 3 | 235 | 30395315 | 73 | |
| BF725606.1 | - | 2 - 3 | 233 | 73 | ||
| BQ694106.1 | - | 2 - 3 | 264 | 73 | ||
| CR541931.1 | - | 1 - 2 | 80 | 73 | ||
| CR541953.1 | - | 1 - 2 | 80 | 73 | ||
| D11428.1 | - | 2 - 3 | 268 | 73 | ||
| DB504971.1 | - | 2 - 3 | 319 | 74 | ||
| HY119295.1 | - | 1 - 2 | 197 | 73 | ||
| KR259963.1 | - | 2 - 3 | 287 | 73 | ||
| KR259964.1 | - | 2 - 3 | 170 | 73 | ||
| L03203.1 | - | 2 - 3 | 194 | 73 | ||
| M94048.1 | - | 1 - 2 | 129 | 73 | ||
More Information
Set Membership: |
|

PMP22
Genomic Map