Assay Details
| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000304.3 | NP_000295.1 | 3 - 4 | 415 | 61 | |
| NM_001281455.1 | NP_001268384.1 | 3 - 4 | 376 | 61 | ||
| NM_001281456.1 | NP_001268385.1 | 3 - 4 | 411 | 61 | ||
| NM_153321.2 | NP_696996.1 | 3 - 4 | 372 | 61 | ||
| NM_153322.2 | NP_696997.1 | 2 - 3 | 294 | 61 | ||
| NR_104017.1 | 2 - 3 | 303 | 61 | |||
| XM_017024775.1 | XP_016880264.1 | 2 - 3 | 211 | 61 | ||
| XM_017024776.1 | XP_016880265.1 | 3 - 4 | 384 | 61 | ||
| GenBank mRNA | AK290640.1 | - | 3 - 4 | 325 | 61 | |
| AK300690.1 | - | 3 - 4 | 267 | 61 | ||
| BC019040.2 | - | 3 - 4 | 337 | 4588473 | 61 | |
| BC091499.1 | - | 3 - 4 | 332 | 30395315 | 61 | |
| BF725606.1 | - | 3 - 4 | 330 | 61 | ||
| BQ232542.1 | - | 1 - 2 | 84 | 61 | ||
| BQ694106.1 | - | 3 - 4 | 361 | 61 | ||
| BX464118.2 | - | 3 - 4 | 370 | 61 | ||
| CR541931.1 | - | 2 - 3 | 177 | 61 | ||
| CR541953.1 | - | 2 - 3 | 177 | 61 | ||
| D11428.1 | - | 3 - 4 | 365 | 61 | ||
| DB504971.1 | - | 3 - 4 | 416 | 61 | ||
| HY119295.1 | - | 2 - 3 | 294 | 61 | ||
| KR259963.1 | - | 3 - 4 | 384 | 61 | ||
| KR259964.1 | - | 3 - 4 | 267 | 61 | ||
| L03203.1 | - | 3 - 4 | 291 | 61 | ||
| M94048.1 | - | 2 - 3 | 226 | 61 | ||
More Information
Set Membership: |
|

PMP22
Genomic Map