Assay Details
| Gene Symbol: DEFB125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153325.3 | NP_697020.2 | 1 - 2 | 94 | 89 | |
| GenBank mRNA | AF525927.1 | - | 1 - 2 | 56 | 89 | |
| AK311802.1 | - | 1 - 2 | 94 | 89 | ||
| AY122477.1 | - | 1 - 2 | 44 | 89 | ||
| BC128195.1 | - | 1 - 2 | 56 | 40113902 | 89 | |
| BC128196.1 | - | 1 - 2 | 56 | 40113904 | 89 | |
More Information
Set Membership: |
|

DEFB125
Genomic Map