Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178100.1 | NP_001171571.1 | 1 - 2 | 102 | 94 | |
| NM_004931.4 | NP_004922.1 | 1 - 2 | 102 | 94 | ||
| NM_172101.3 | NP_742099.1 | 1 - 2 | 102 | 94 | ||
| NM_172102.3 | NP_742100.1 | 1 - 2 | 102 | 94 | ||
| NM_172213.3 | NP_757362.1 | 1 - 2 | 102 | 94 | ||
| XM_011533164.2 | XP_011531466.1 | 1 - 2 | 60 | 94 | ||
| GenBank mRNA | BC100912.2 | - | 1 - 2 | 93 | 40003723 | 94 |
More Information
Set Membership: |
|

CD8B
Genomic Map