Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172102.3 | NP_742100.1 | 3 - 4 | 551 | 89 | |
| GenBank mRNA | X13446.1 | - | 3 - 4 | 542 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172102.3 | NP_742100.1 | 3 - 4 | 551 | 89 | |
| GenBank mRNA | X13446.1 | - | 3 - 4 | 542 | 89 | |
Set Membership: |
|