Assay Details
| Gene Symbol: C11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173525.2 | NP_775796.2 | 2 - 3 | 917 | 87 | |
| GenBank mRNA | BC031612.1 | - | 2 - 3 | 917 | 5171107 | 87 |
More Information
Set Membership: |
|

C11orf42
Genomic Map