Assay Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003712.3 | NP_003703.1 | 5 - 6 | 814 | 87 | |
| NM_177526.2 | NP_803545.1 | 5 - 6 | 773 | 87 | ||
| NM_177543.2 | NP_808211.1 | 5 - 6 | 881 | 87 | ||
| XM_011528396.2 | XP_011526698.1 | 5 - 6 | 810 | 87 | ||
| GenBank mRNA | AB209696.1 | - | 2 - 3 | 5962 | 87 | |
| AF035959.1 | - | 5 - 6 | 758 | 87 | ||
| AF047760.1 | - | 5 - 6 | 758 | 87 | ||
| AF056083.1 | - | 5 - 6 | 758 | 87 | ||
| BC002806.2 | - | 5 - 6 | 765 | 3659728 | 87 | |
| BM982485.1 | - | 1 - 2 | 511 | 87 | ||
| BT007021.1 | - | 5 - 6 | 715 | 87 | ||
More Information
Set Membership: |
|

PLPP2
Genomic Map