Assay Details
| Gene Symbol: CMTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181271.2 | NP_851788.1 | 1 - 2 | 217 | 114 | |
| GenBank mRNA | AY174120.1 | - | 1 - 2 | 221 | 114 | |
More Information
Set Membership: |
|

CMTM1
Genomic Map