Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242416.1 | NP_001229345.1 | 1 - 2 | 318 | 75 | |
| NM_181308.2 | NP_851825.1 | 1 - 2 | 318 | 75 | ||
| XM_006720308.1 | XP_006720371.1 | 1 - 2 | 310 | 75 | ||
| GenBank mRNA | AK225852.1 | - | 1 - 2 | 277 | 75 | |
| BX248274.1 | - | 1 - 2 | 220 | 75 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map