Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181460.3 | NP_852125.1 | 7 - 8 | 1555 | 105 | |
| NM_181461.3 | NP_852126.1 | 7 - 8 | 1555 | 276 | ||
| GenBank mRNA | AY251280.1 | - | 7 - 8 | 1225 | 105 | |
More Information
Set Membership: |
|

PAX3
Genomic Map