Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181460.3 | NP_852125.1 | 8 - 9 | 1588 | 76 | |
| GenBank mRNA | AY251280.1 | - | 8 - 9 | 1258 | 76 | |
More Information
Set Membership: |
|

PAX3
Genomic Map