Assay Details
| Gene Symbol: GDPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182833.1 | NP_878253.1 | 8 - 9 | 882 | 106 | |
| XM_011544826.1 | XP_011543128.1 | 8 - 9 | 2613 | 106 | ||
| XM_011544833.1 | XP_011543135.1 | 8 - 9 | 2613 | 106 | ||
| XM_011544834.1 | XP_011543136.1 | 9 - 10 | 1108 | 106 | ||
| XM_017017348.1 | XP_016872837.1 | 9 - 10 | 1052 | 106 | ||
| XM_017017349.1 | XP_016872838.1 | 8 - 9 | 969 | 106 | ||
| XM_017017350.1 | XP_016872839.1 | 8 - 9 | 2484 | 106 | ||
| XM_017017351.1 | XP_016872840.1 | 8 - 9 | 840 | 106 | ||
| GenBank mRNA | AY313782.1 | - | 8 - 9 | 882 | 106 | |
| AY326450.1 | - | 8 - 9 | 882 | 106 | ||
More Information
Set Membership: |
|

GDPD4
Genomic Map