Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197958.2 | NP_932062.1 | 1 - 2 | 373 | 74 | |
| GenBank mRNA | BC039153.1 | - | 1 - 2 | 229 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LARP6| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197958.2 | NP_932062.1 | 1 - 2 | 373 | 74 | |
| GenBank mRNA | BC039153.1 | - | 1 - 2 | 229 | 74 | |
Set Membership: |
|