Assay Details
| Gene Symbol: FAM120C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198456.2 | NP_940858.2 | 1 - 2 | 781 | 70 | |
| GenBank mRNA | AK000513.1 | - | 1 - 2 | 725 | 70 | |
| AY121803.1 | - | 1 - 2 | 781 | 70 | ||
| BC016138.1 | - | 1 - 2 | 781 | 70 | ||
| BC081532.1 | - | 1 - 2 | 482 | 70 | ||
More Information
Set Membership: |
|

FAM120C
Genomic Map