Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206824.2 | NP_996560.1 | 1 - 2 | 443 | 79 | |
| GenBank mRNA | AK129513.1 | - | 1 - 2 | 241 | 79 | |
| AK289790.1 | - | 1 - 2 | 342 | 79 | ||
| CD249837.1 | - | 244 | 79 | |||
| KU178763.1 | - | 1 - 2 | 173 | 79 | ||
More Information
Set Membership: |
|

VKORC1
Genomic Map