Assay Details
| Gene Symbol: TOMM20L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207377.2 | NP_997260.1 | 2 - 3 | 214 | 104 | |
| GenBank mRNA | AY358266.1 | - | 2 - 3 | 226 | 104 | |
| BC137557.1 | - | 2 - 3 | 212 | 9021557 | 104 | |
| BC137563.1 | - | 2 - 3 | 212 | 9021563 | 104 | |
More Information
Set Membership: |
|

TOMM20L
Genomic Map