Assay Details
| Gene Symbol: FAM19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252216.1 | NP_001239145.1 | 3 - 4 | 470 | 69 | |
| NM_213609.3 | NP_998774.2 | 3 - 4 | 744 | 69 | ||
| XM_017006386.1 | XP_016861875.1 | 2 - 3 | 891 | 69 | ||
| XM_017006387.1 | XP_016861876.1 | 3 - 4 | 1383 | 69 | ||
| XM_017006388.1 | XP_016861877.1 | 3 - 4 | 1176 | 69 | ||
| XM_017006389.1 | XP_016861878.1 | 2 - 3 | 888 | 69 | ||
| XM_017006390.1 | XP_016861879.1 | 3 - 4 | 1384 | 69 | ||
| GenBank mRNA | AK289668.1 | - | 3 - 4 | 733 | 69 | |
| AK290446.1 | - | 3 - 4 | 666 | 69 | ||
| AY325114.1 | - | 3 - 4 | 470 | 69 | ||
| BC025746.1 | - | 3 - 4 | 646 | 5200142 | 69 | |
More Information
Set Membership: |
|

FAM19A1
Genomic Map