Assay Details
| Gene Symbol: ECHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105545.1 | NP_001099015.1 | 2 - 3 | 411 | 76 | |
| NM_018479.3 | NP_060949.2 | 3 - 4 | 633 | 76 | ||
| XM_005267049.2 | XP_005267106.1 | 3 - 4 | 900 | 76 | ||
| XM_005267050.2 | XP_005267107.1 | 4 - 5 | 526 | 76 | ||
| GenBank mRNA | BQ224769.1 | - | 3 - 4 | 579 | 76 | |
| DA846978.1 | - | 2 - 3 | 351 | 76 | ||
| EU668355.1 | - | 2 - 3 | 411 | 76 | ||
More Information
Set Membership: |
|

ECHDC1
Genomic Map